3-Brom-N-methylbenzolsulfonamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR5804-1G
OR5804-5G
APOSOR5804-1GEA
176
EUR
APOSOR5804-1G
APOSOR5804-5G
3-Brom-N-methylbenzolsulfonamid 98%
3-Brom-N-methylbenzolsulfonamid
Formel:
C₇H₈BrNO₂S Molecular Weight: 250,11 g/mol Schmelzpunkt: 62…66 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363818 CAS-Nummer: 153435-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...