1-((3-Bromphenyl)sulfonyl)piperidin 98%
Lieferant: Apollo Scientific
OR5805-1G
OR5805-25G
OR5805-5G
APOSOR5805-1GEA
121
EUR
APOSOR5805-1G
APOSOR5805-25G
APOSOR5805-5G
1-((3-Bromphenyl)sulfonyl)piperidin 98%
1-((3-Bromphenyl)sulfonyl)piperidin
Formel:
C₁₁H₁₄BrNO₂S Molecular Weight: 304,2000 g/mol Schmelzpunkt: 80…84 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363827 CAS-Nummer: 871269-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...