2,5-Dimethoxyphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,5-Dimethoxyphenyl)metaborsäure, 2,5-Dimethoxybenzolboronsäure, Dihydroxy(2,5-dimethoxyphenyl)boran

OR5823-100G
APOSOR5823-100GEA 197 EUR
APOSOR5823-100G
2,5-Dimethoxyphenylboronsäure
2,5-Dimethoxyphenylboronsäure
Formel: C₈H₁₁BO₄
Molecular Weight: 181,98 g/mol
Schmelzpunkt: 95…98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318181
CAS-Nummer: 107099-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR