2-Amino-6-methoxy-3-nitropyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-Methoxy-3-nitropyridin-2-amine
OR5827-100G
OR5827-25G
OR5827-5G
APOSOR5827-100GEA
145
EUR
APOSOR5827-100G
APOSOR5827-25G
APOSOR5827-5G
2-Amino-6-methoxy-3-nitropyridin
2-Amino-6-methoxy-3-nitropyridine
Formel:
C₆H₇N₃O₃ Molecular Weight: 169,14 g/mol Schmelzpunkt: 170…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03206481 CAS-Nummer: 73896-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...