6-Methyl-3(2H)-pyridazinon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methylpyridazin-3(2H)-one

OR5828-100G OR5828-25G OR5828-500G
APOSOR5828-100GEA 394 EUR
APOSOR5828-100G APOSOR5828-25G APOSOR5828-500G
6-Methyl-3(2H)-pyridazinon 97%
6-Methyl-3(2H)-pyridazinon
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039720
CAS-Nummer: 13327-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR