Xanthane hydride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3H-1,2,4-dithiazole-3-thione

OR5839-100G OR5839-25G OR5839-500G
APOSOR5839-100GEA 213 EUR
APOSOR5839-100G APOSOR5839-25G APOSOR5839-500G
Xanthane hydride 95%
Xanthane hydride
Formel: C₂H₂N₂S₃
Molecular Weight: 150,23 g/mol
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051660
CAS-Nummer: 6846-35-1
EINECS: 229-933-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR