Pyrimidin-2-ol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Hydroxypyrimidine
OR5850-5G
OR5850-1G
APOSOR5850-5GEA
90.2
EUR
APOSOR5850-5G
APOSOR5850-1G
Pyrimidin-2-ol 95+%
Pyrimidin-2-ol
Formel:
C4H4N2O Molecular Weight: 96,0900 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047358 CAS-Nummer: 51953-13-0 EINECS: 209-149-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...