Ethyl-3-oxo-3-(4-pyridinyl)propanoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-oxo-3-(pyridin-4-yl)propanoate, Ethyl isonicotinoylacetate, Ethyl 3-oxo-3-(4-pyridyl)propionate

OR5896-1G OR5896-5G
APOSOR5896-1GEA 21.9 EUR
APOSOR5896-1G APOSOR5896-5G
Ethyl-3-oxo-3-(4-pyridinyl)propanoat
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,20 g/mol
Schmelzpunkt: 53…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00094021
CAS-Nummer: 26377-17-3
EINECS: 247-635-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR