Ethylpicolinoylacetat 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-oxo-3-(2-pyridyl)propionate, Ethyl 3-oxo-3-(pyridin-2-yl)propanoate

OR5897-25G OR5897-500MG OR5897-5G
APOSOR5897-25GEA 426 EUR
APOSOR5897-25G APOSOR5897-500MG APOSOR5897-5G
Ethylpicolinoylacetat 95+%
Ethylpicolinoylacetat
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,20 g/mol
Siedepunkt: 121…123 °C/0,6mmHg
Dichte: 1,164 g/cm³ (25 °C)
Flammpunkt: >200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00094022
CAS-Nummer: 26510-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR