4-Methylbenzamidoxim 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Hydroxy-4-methylbenzimidamide

OR5904-1G OR5904-25G OR5904-5G
APOSOR5904-1GEA 36.4 EUR
APOSOR5904-1G APOSOR5904-25G APOSOR5904-5G
4-Methylbenzamidoxim 97%
4-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
Schmelzpunkt: 140…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019952
CAS-Nummer: 19227-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR