(+/-)-3-(Boc-aminomethyl)piperidine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Aminomethyl)piperidine, 3-BOC protected

OR5910-5G OR5910-1G OR5910-25G
APOSOR5910-5GEA 49.3 EUR
APOSOR5910-5G APOSOR5910-1G APOSOR5910-25G
(+/-)-3-(Boc-aminomethyl)piperidine 97%
(+/-)-3-(Boc-aminomethyl)piperidine
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,309 g/mol
Schmelzpunkt: 60…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01632454
CAS-Nummer: 142643-29-6
UN: 3259
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR