[2-(Morpholin-4-ylcarbonyl)phenyl]amine

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Aminophenyl)(morpholin-4-yl)methanone

OR5921-1G OR5921-250MG
APOSOR5921-1GEA 34.8 EUR
APOSOR5921-1G APOSOR5921-250MG
[2-(Morpholin-4-ylcarbonyl)phenyl]amine
[2-(Morpholin-4-ylcarbonyl)phenyl]amine
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,25 g/mol
Schmelzpunkt: 74…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00034763
CAS-Nummer: 39630-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR