5-Amino-2-methylindol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole

OR59321-100G OR59321-250G OR59321-25G OR59321-5G
APOSOR59321-100GEA 219 EUR
APOSOR59321-100G APOSOR59321-250G APOSOR59321-25G APOSOR59321-5G
5-Amino-2-methylindol 95%
5-Amino-2-methylindole
Formel: C₉H₁₀N₂
Molecular Weight: 146.19 g/mol
Schmelzpunkt: 153…157 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093426
CAS-Nummer: 7570-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR