2-Amino-5-brompyrimidin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Brompyrimidin-2-ylamin
OR5933-100G
OR5933-25G
OR5933-5G
APOSOR5933-100GEA
55.1
EUR
APOSOR5933-100G
APOSOR5933-25G
APOSOR5933-5G
2-Amino-5-brompyrimidin
2-Amino-5-bromopyrimidine
Formel:
C₄H₄BrN₃ Molecular Weight: 174 g/mol Schmelzpunkt: 241…243 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012341 CAS-Nummer: 7752-82-1 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...