2-Chlor-6-(dipropylamino)pyrazin
Lieferant: Apollo Scientific
OR59332-1G
OR59332-5G
APOSOR59332-1GEA
132
EUR
APOSOR59332-1G
APOSOR59332-5G
2-Chlor-6-(dipropylamino)pyrazin
2-Chloro-6-(dipropylamino)pyrazine
Formel:
C₁₀H₁₆ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878390 CAS-Nummer: 957065-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...