2-Bromisobuttersäureamid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-2-methylpropanamide

OR59336-25G OR59336-5G
APOSOR59336-25GEA 190 EUR
APOSOR59336-25G APOSOR59336-5G
2-Bromisobuttersäureamid
2-Bromisobuttersäureamid
Formel: C₄H₈BrNO
Molecular Weight: 166.0164 g/mol
Schmelzpunkt: 145…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08234837
CAS-Nummer: 7462-74-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR