2-Acetamido-5-chlorphenylboronsäurepinakolester
Lieferant: Apollo Scientific
OR59347-1G
OR59347-5G
APOSOR59347-1GEA
190
EUR
APOSOR59347-1G
APOSOR59347-5G
2-Acetamido-5-chlorphenylboronsäurepinakolester
2-Acetamido-5-chlorphenylboronsäurepinakolester
Formel:
C₁₄H₁₉BClNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878425 CAS-Nummer: 957063-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...