Methyl-4-chlorsalicylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-chloro-2-hydroxybenzoate

OR59351-25G
APOSOR59351-25GEA 78.3 EUR
APOSOR59351-25G
Methyl-4-chlorsalicylat
Methyl-4-chlorsalicylat
Formel: C₈H₇ClO₃
Molecular Weight: 186,5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407436
CAS-Nummer: 22717-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR