8-Brom-6-methyl-[1,2,4]triazolo[1,5-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Bromo-6-methyl-[1,2,4]triazolo[1,5-a]pyridine
OR59356-1G
OR59356-500MG
APOSOR59356-1GEA
62.5
EUR
APOSOR59356-1G
APOSOR59356-500MG
8-Brom-6-methyl-[1,2,4]triazolo[1,5-a]pyridin
8-Brom-6-methyl-[1,2,4]triazolo[1,5-a]pyridin
Formel:
C₇H₆BrN₃ Molecular Weight: 212,0500 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09878402 CAS-Nummer: 957062-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...