6-Chloro-N-isopropyl-4-pyrimidinamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-N-isopropylpyrimidin-4-amine, 4-Chloro-6-(isopropylamino)pyrimidine
OR59375-1G
OR59375-5G
APOSOR59375-1GEA
94.8
EUR
APOSOR59375-1G
APOSOR59375-5G
6-Chloro-N-isopropyl-4-pyrimidinamine
6-Chloro-N-isopropyl-4-pyrimidinamine
Formel:
C₇H₁₀ClN₃ Molecular Weight: 171.63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09756956 CAS-Nummer: 945896-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...