5-(2,4-Dichlorphenyl)-4-(hydroxymethyl)oxazol
Lieferant: Apollo Scientific
OR59376-1G
OR59376-250MG
APOSOR59376-1GEA
430
EUR
APOSOR59376-1G
APOSOR59376-250MG
5-(2,4-Dichlorphenyl)-4-(hydroxymethyl)oxazol
5-(2,4-Dichlorophenyl)-4-(hydroxymethyl)oxazole
Formel:
C₁₀H₇Cl₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878381 CAS-Nummer: 957062-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...