N-Benzyl-2-brombenzolsulfonamid

Lieferant: Apollo Scientific

OR59379-1G OR59379-5G
APOSOR59379-1GEA 132 EUR
APOSOR59379-1G APOSOR59379-5G
N-Benzyl-2-brombenzolsulfonamid
N-Benzyl-2-brombenzolsulfonamid
Formel: C₁₃H₁₂BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD02555935
CAS-Nummer: 321704-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR