1-(3-Chlor-4-methylphenyl)piperidin

Lieferant: Apollo Scientific

OR59388-1G OR59388-5G
APOSOR59388-1GEA 132 EUR
APOSOR59388-1G APOSOR59388-5G
1-(3-Chlor-4-methylphenyl)piperidin
1-(3-Chlor-4-methylphenyl)piperidin
Formel: C₁₂H₁₆ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD09878387
CAS-Nummer: 1000339-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR