1-(3-Chlorphenyl)pyrrolidin

Lieferant: Apollo Scientific

OR59390-1G OR59390-5G
APOSOR59390-1GEA 125 EUR
APOSOR59390-1G APOSOR59390-5G
1-(3-Chlorphenyl)pyrrolidin
1-(3-Chlorphenyl)pyrrolidin
Formel: C₁₀H₁₂ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD09878398
CAS-Nummer: 88154-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR