4'-Brom-3'-chlor-2'-methylacetanilid

Lieferant: Apollo Scientific

Synonyme: N-(4-Bromo-3-chloro-2-methylphenyl)acetamide

OR59398-1G OR59398-5G
APOSOR59398-1GEA 30.6 EUR
APOSOR59398-1G APOSOR59398-5G
4'-Brom-3'-chlor-2'-methylacetanilid
4'-Brom-3'-chlor-2'-methylacetanilid
Formel: C₉H₉BrClNO
Molecular Weight: 262,5300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059123
CAS-Nummer: 125328-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR