4'-Brom-2'-hydroxyacetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Brom-2-hydroxyphenyl)ethan-1-on, 2-Acetyl-5-bromphenol

OR59425-1G OR59425-5G
APOSOR59425-1GEA 21.9 EUR
APOSOR59425-1G APOSOR59425-5G
4'-Brom-2'-hydroxyacetophenon 98%
4'-Brom-2'-hydroxyacetophenon
Formel: C₈H₇BrO₂
Molecular Weight: 215.05
Lagertemperatur: Raumtemperatur
MDL: MFCD03428533
CAS-Nummer: 30186-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR