2-Brom-5-nitroanilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR59426-25G
OR59426-5G
OR59426-100G
APOSOR59426-25GEA
45.1
EUR
APOSOR59426-25G
APOSOR59426-5G
APOSOR59426-100G
2-Brom-5-nitroanilin 98%
2-Brom-5-nitroanilin
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217,02 g/mol Schmelzpunkt: 137…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051578 CAS-Nummer: 10403-47-1 EINECS: 233-874-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...