5-Chlor-2-pyrazincarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR59462-1G
OR59462-5G
OR59462-10G
APOSOR59462-1GEA
21.9
EUR
APOSOR59462-1G
APOSOR59462-5G
APOSOR59462-10G
5-Chlor-2-pyrazincarbonsäure
5-Chloropyrazine-2-carboxylic acid
Formel:
C5H3ClN2O2 Molecular Weight: 158,54 g/mol Siedepunkt: 330 °C Schmelzpunkt: 150…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033269 CAS-Nummer: 36070-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...