6-Chlor-2-pyridinboronsäurepinakolester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Chloropyridine-2-boronic acid pinacol ester
OR59467-250MG
APOSOR59467-250MGEA
21.9
EUR
APOSOR59467-250MG
6-Chlor-2-pyridinboronsäurepinakolester 98%
6-Chlor-2-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₅BClNO₂ Molecular Weight: 239,51 g/mol Schmelzpunkt: 109…111 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD06798259 CAS-Nummer: 652148-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...