5-Methyl-thiophene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methylthiophene-3-carboxylic acid
OR5951-1G
OR5951-250MG
OR5951-5G
APOSOR5951-1GEA
52.2
EUR
APOSOR5951-1G
APOSOR5951-250MG
APOSOR5951-5G
5-Methyl-thiophene-3-carboxylic acid
5-Methyl-thiophene-3-carboxylic acid
Formel:
C₆H₆O₂S Molecular Weight: 142.18 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01680229 CAS-Nummer: 19156-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...