4'-Brom-3-chlorpropiophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Brom-β-chlorpropiophenon

OR5955-100G
APOSOR5955-100GEA 177 EUR
APOSOR5955-100G
4'-Brom-3-chlorpropiophenon 98%
4'-Brom-3-chlorpropiophenon
Formel: C₉H₈BrClO
Molecular Weight: 247,52 g/mol
Schmelzpunkt: 54…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00018979
CAS-Nummer: 31736-73-9
EINECS: 250-784-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR