4-Acetoxybenzeneboronic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Acetoxybenzeneboronic acid, 4-Acetoxyphenylboronic acid, (4-Acetoxyphenyl)boronic acid
OR5971-1G
OR5971-5G
APOSOR5971-1GEA
102
EUR
APOSOR5971-1G
APOSOR5971-5G
4-Acetoxybenzeneboronic acid 97%
4-Acetoxybenzeneboronic acid
Formel:
C₈H₉BO₄ Molecular Weight: 179.97 g/mol Schmelzpunkt: 220…225 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09027198 CAS-Nummer: 177490-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...