Methyl-2-hydroxy-5-nitronicotinat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-nitro-2-oxo-1,2-dihydropyridine-3-carboxylate
OR59785-1G
OR59785-5G
APOSOR59785-1GEA
94.5
EUR
APOSOR59785-1G
APOSOR59785-5G
Methyl-2-hydroxy-5-nitronicotinat 98%
Methyl-2-hydroxy-5-nitronicotinat
Formel:
C₇H₆N₂O₅ Molecular Weight: 198,14 g/mol Schmelzpunkt: 250…254 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07783889 CAS-Nummer: 856579-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...