3-Iodo-4-pyridine carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Iodoisonicotinic acid, 3-Iodopyridine-4-carboxylic acid
OR59795-1G
OR59795-5G
OR59795-250MG
APOSOR59795-1GEA
183
EUR
APOSOR59795-1G
APOSOR59795-5G
APOSOR59795-250MG
3-Iodo-4-pyridine carboxylic acid
3-Iodo-4-pyridine carboxylic acid
Formel:
C₆H₄INO₂ Molecular Weight: 249.01 g/mol Schmelzpunkt: 241…243 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646072 CAS-Nummer: 57842-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...