2-(Methylthio)pyrimidin-4-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2-(methylthio)pyrimidine

OR59797-100MG OR59797-1G
APOSOR59797-100MGEA 21.9 EUR
APOSOR59797-100MG APOSOR59797-1G
2-(Methylthio)pyrimidin-4-amine
2-(Methylthio)pyrimidin-4-amine
Formel: C₅H₇N₃S
Molecular Weight: 141,1942 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02091098
CAS-Nummer: 2183-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR