2-Amino-3-brompyridin 98%

Lieferant: Apollo Scientific
Achtung

OR59810-25G OR59810-10G OR59810-5G
APOSOR59810-25GEA 75.4 EUR
APOSOR59810-25G APOSOR59810-10G APOSOR59810-5G
2-Amino-3-brompyridin 98%
2-Amino-3-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Schmelzpunkt: 63…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095187
CAS-Nummer: 13534-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR