2-Amino-4,5-dichlorpyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4,5-Dichloropyridin-2-amine
OR59818-1G
OR59818-250MG
OR59818-5G
APOSOR59818-1GEA
29
EUR
APOSOR59818-1G
APOSOR59818-250MG
APOSOR59818-5G
2-Amino-4,5-dichlorpyridin
2-Amino-4,5-dichloropyridine
Formel:
C₅H₄Cl₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437842 CAS-Nummer: 188577-68-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...