Chroman-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR59826-1G OR59826-250MG
APOSOR59826-1GEA 437 EUR
APOSOR59826-1G APOSOR59826-250MG
Chroman-6-carboxylic acid
Chroman-6-carboxylic acid
Formel: C₁₀H₁₀O₃
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08146590
CAS-Nummer: 103203-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR