5-Brom-2-nitrobenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR59833-1G
OR59833-25G
OR59833-5G
APOSOR59833-1GEA
21.9
EUR
APOSOR59833-1G
APOSOR59833-25G
APOSOR59833-5G
5-Brom-2-nitrobenzoesäure
5-Brom-2-nitrobenzoesäure
Formel:
C₇H₄BrNO₄ Molecular Weight: 246,02 g/mol Schmelzpunkt: 139…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093013 CAS-Nummer: 6950-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...