2-(Hydroxymethyl)-1,3-oxazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Oxazolemethanol
OR59845-1G
OR59845-250MG
APOSOR59845-1GEA
253
EUR
APOSOR59845-1G
APOSOR59845-250MG
2-(Hydroxymethyl)-1,3-oxazole
2-(Hydroxymethyl)-1,3-oxazole
Formel:
C₄H₅NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08703642 CAS-Nummer: 130551-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...