2-Amino-5-pyrimidinboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Aminopyrimidine-5-boronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 2-(2-Aminopyrimidin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR59880-10G OR59880-25G
APOSOR59880-10GEA 85.7 EUR
APOSOR59880-10G APOSOR59880-25G
2-Amino-5-pyrimidinboronsäurepinakolester 98%
2-Amino-5-pyrimidinboronsäurepinakolester
Formel: C₁₀H₁₆BN₃O₂
Molecular Weight: 221,06 g/mol
Schmelzpunkt: 209…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795685
CAS-Nummer: 402960-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR