2-Amino-4-chlorothiazole-5-carbaldehyde 98%

Lieferant: Apollo Scientific
Achtung

OR59894-1G OR59894-5G OR59894-10G
APOSOR59894-1GEA 33.5 EUR
APOSOR59894-1G APOSOR59894-5G APOSOR59894-10G
2-Amino-4-chlorothiazole-5-carbaldehyde 98%
2-Amino-4-chlorothiazole-5-carbaldehyde
Formel: C₄H₃ClN₂OS
Molecular Weight: 162.5974 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00204993
CAS-Nummer: 76874-79-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR