(3-Chlorphenyl)essigsäure 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chlorophenylacetic acid

OR5991-100G OR5991-25G OR5991-500G
APOSOR5991-100GEA 42.2 EUR
APOSOR5991-100G APOSOR5991-25G APOSOR5991-500G
(3-Chlorphenyl)essigsäure 98+%
(3-Chlorphenyl)essigsäure
Formel: (C₆H₄Cl)CH₂COOH
Molecular Weight: 170,6 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004332
CAS-Nummer: 1878-65-5
EINECS: 217-520-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR