(4-Biphenyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Biphenylacetic acid
OR5992-1KG
OR5992-250G
OR5992-5KG
APOSOR5992-1KGEA
154
EUR
APOSOR5992-1KG
APOSOR5992-250G
APOSOR5992-5KG
(4-Biphenyl)essigsäure 97%
(4-Biphenyl)essigsäure
Formel:
(C₆H₅C₆H₅)CH₂COOH Molecular Weight: 212,25 g/mol Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004351 CAS-Nummer: 5728-52-9 EINECS: 227-233-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...