trans-2-Phenylcyclopropancarbonsäure
Lieferant: Apollo Scientific
Synonyme:
trans-2-Phenylcyclopropane-1-carboxylic acid
OR59934-25G
APOSOR59934-25GEA
119
EUR
APOSOR59934-25G
trans-2-Phenylcyclopropancarbonsäure
trans-2-Phenylcyclopropanecarboxylic acid
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,1852 g/mol Schmelzpunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001292 CAS-Nummer: 939-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...