5-Pyridin-4-ylthiophene-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR59948-1G OR59948-5G
APOSOR59948-1GEA 306 EUR
APOSOR59948-1G APOSOR59948-5G
5-Pyridin-4-ylthiophene-2-carboxylic acid 95%
5-Pyridin-4-ylthiophene-2-carboxylic acid
Formel: C₁₀H₇NO₂S
Molecular Weight: 205.24 g/mol
Schmelzpunkt: 333.5…337 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07376007
CAS-Nummer: 216867-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR