(2,3-Dichlorphenyl)essigsäure

Lieferant: Apollo Scientific

Synonyme: 2,3-Dichlorophenylacetic acid

OR59959-100G OR59959-25G
APOSOR59959-100GEA 116 EUR
APOSOR59959-100G APOSOR59959-25G
(2,3-Dichlorphenyl)essigsäure
(2,3-Dichlorphenyl)essigsäure
Formel: (C₆H₃Cl₂)CH₂COOH
Molecular Weight: 205.04 g/mol
Schmelzpunkt: 130…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861393
CAS-Nummer: 10236-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR