2-Amino-4-phenylphenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-4-hydroxybiphenyl, 3-Amino-[1,1'-biphenyl]-4-ol
OR60002-1G
OR60002-250MG
APOSOR60002-1GEA
33.5
EUR
APOSOR60002-1G
APOSOR60002-250MG
2-Amino-4-phenylphenol 98%
2-Amino-4-phenylphenol
Formel:
C₁₂H₁₁NO Molecular Weight: 185,2200 g/mol Schmelzpunkt: 198…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059187 CAS-Nummer: 1134-36-7 EINECS: 214-484-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...