1-Methyl-2-benzimidazolinone 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dihydro-1-methyl-2H-benzimidazol-2-one, 1-Methyl-1H-benzo[d]imidazol-2(3H)-one
OR60004-1G
OR60004-25G
OR60004-5G
APOSOR60004-1GEA
37.7
EUR
APOSOR60004-1G
APOSOR60004-25G
APOSOR60004-5G
1-Methyl-2-benzimidazolinone 96%
1-Methyl-2-benzimidazolinone
Formel:
C₈H₈N₂O Molecular Weight: 148.165 g/mol Schmelzpunkt: 192…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142654 CAS-Nummer: 1849-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...