2-Amino-4,6-dichlorpyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR60009-1G
OR60009-5G
OR60009-10G
APOSOR60009-1GEA
21.9
EUR
APOSOR60009-1G
APOSOR60009-5G
APOSOR60009-10G
2-Amino-4,6-dichlorpyridin
2-Amino-4,6-dichloropyridine
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,0100 g/mol Schmelzpunkt: 108…113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04037218 CAS-Nummer: 116632-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...